siRNA Details
Gene Symbol: WDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017491.3 | NP_059830.1 | 2, 3 | 419 | ||
XM_017008880.1 | XP_016864369.1 | 2, 3 | 453 | |||
GenBank mRNA | AB010427.2 | - | 2, 3 | 338 | ||
AB209746.1 | - | 2, 3 | 192 | |||
AF020056.1 | - | 2, 3 | 247 | |||
AF020260.1 | - | 2, 3 | 247 | |||
AK127665.1 | - | 2, 3 | 264 | |||
AK222764.1 | - | 2, 3 | 291 | |||
AK222897.1 | - | 2, 3 | 323 | |||
AK225276.1 | - | 2, 3 | 291 | |||
AK291614.1 | - | 2, 3 | 269 | |||
AK299602.1 | - | 2, 3 | 264 | |||
BC000201.1 | - | 2, 3 | 218 | 3351638 | ||
BC002489.2 | - | 2, 3 | 210 | 3051319 | ||
BI831941.1 | - | 2, 3 | 420 | |||
EU794675.1 | - | 2, 3 | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
