siRNA Details
Gene Symbol: GOLGA8F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033351.1 | 18 | 2157 | |||
GenBank mRNA | AK057241.1 | - | 16 | 1635 | ||
AK302625.1 | - | 16 | 1582 | |||
AL833871.1 | - | 7 | 637 | |||
BC144672.1 | - | 15 | 1762 | |||
BC150302.1 | - | 18 | 2131 | 8860399 | ||
DQ309038.1 | - | 16 | 1605 |
Gene Symbol: GOLGA8CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027411.2 | 18 | 2169 |
Gene Symbol: GOLGA8DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027407.1 | 18 | 2157 | |||
GenBank mRNA | BC110417.1 | - | 18 | 2128 | 5171794 |
Gene Symbol: GOLGA8EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033350.1 | 18 | 2157 | |||
GenBank mRNA | BC063309.1 | - | 19 | 2131 | ||
DQ309037.1 | - | 18 | 2131 |
Gene Symbol: GOLGA8G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033353.1 | 18 | 2169 | |||
GenBank mRNA | BC066774.1 | - | 18 | 2143 | ||
BC099913.1 | - | 17 | 2984 | 4838463 | ||
BC125193.1 | - | 17 | 1949 | 40122746 | ||
BC125194.1 | - | 17 | 1949 | 40122749 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
