siRNA Details
Gene Symbol: ZNF137P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023311.1 | 1 | 72 | |||
GenBank mRNA | BC110455.1 | - | 1 | 72 | ||
BC110456.1 | - | 1 | 72 | |||
U09414.1 | - | 1 | 106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
