siRNA Details
Gene Symbol: ZNF761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008401.3 | NP_001008401.3 | 6 | 3330 | ||
NM_001289951.1 | NP_001276880.1 | 5 | 3111 | |||
NM_001289952.1 | NP_001276881.1 | 5 | 3179 | |||
GenBank mRNA | AB107355.2 | - | 5 | 3180 | ||
AK092652.1 | - | 1 | 1664 | |||
AK131281.1 | - | 2 | 2800 |
Gene Symbol: ZNF137P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023311.1 | 1 | 543 | |||
GenBank mRNA | BC110455.1 | - | 1 | 543 | ||
BC110456.1 | - | 1 | 543 | |||
U09414.1 | - | 1 | 577 |
Gene Symbol: ZNF578 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099694.1 | NP_001093164.1 | 6 | 2427 | ||
XM_017026302.1 | XP_016881791.1 | 6 | 2447 | |||
GenBank mRNA | AK055946.1 | - | 1 | 1605 | ||
AK095562.1 | - | 1 | 1122 |
Gene Symbol: ZNF600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321866.1 | NP_001308795.1 | 6 | 3433 | ||
NM_001321867.1 | NP_001308796.1 | 4 | 2810 | |||
NM_198457.3 | NP_940859.3 | 3 | 2683 | |||
GenBank mRNA | BX640933.1 | - | 3 | 2655 |
Gene Symbol: ZNF611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161499.1 | NP_001154971.1 | 6 | 2863 | ||
NM_001161500.1 | NP_001154972.1 | 5 | 2763 | |||
NM_001161501.1 | NP_001154973.1 | 5 | 2726 | |||
NM_030972.3 | NP_112234.3 | 7 | 2905 | |||
GenBank mRNA | AK021787.1 | - | 1 | 648 | ||
AK091389.1 | - | 2 | 2443 | |||
AK097434.1 | - | 7 | 2905 | |||
AL834123.1 | - | 1 | 2267 | |||
BC000918.2 | - | 1 | 298 | |||
BC028590.1 | - | 9 | 3147 |
Gene Symbol: ZNF765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040185.1 | NP_001035275.1 | 4 | 3377 | ||
GenBank mRNA | AK293890.1 | - | 6 | 3593 | ||
BX647981.1 | - | 4 | 3371 |
Gene Symbol: ZNF813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004301.3 | NP_001004301.2 | 4 | 2741 | ||
GenBank mRNA | AK091460.1 | - | 1 | 1889 | ||
AK131422.1 | - | 1 | 2124 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
