siRNA Details
Gene Symbol: SPDYE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716210.3 | XP_006716273.1 | 2 | 862 | ||
XM_017012933.1 | XP_016868422.1 | 2 | 866 |
Gene Symbol: SPDYE17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716212.3 | XP_006716275.1 | 2 | 866 | ||
XM_017012888.1 | XP_016868377.1 | 2 | 866 |
Gene Symbol: SPDYE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004351.4 | NP_001004351.3 | 2 | 492 | ||
XM_011516234.2 | XP_011514536.1 | 2 | 1395 | |||
XM_011516235.2 | XP_011514537.1 | 2 | 1395 | |||
GenBank mRNA | BC056606.1 | - | 2 | 493 | 4838469 | |
BC100972.2 | - | 1 | 293 |
Gene Symbol: SPDYE8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003664.2 | 3 | 903 | |||
GenBank mRNA | BC045636.1 | - | 3 | 906 | ||
BC068610.1 | - | 3 | 906 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
