siRNA Details
Gene Symbol: USP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304284.1 | NP_001291213.1 | 14 | 2572 | ||
NM_004505.3 | NP_004496.2 | 6 | 2041 | |||
XM_011524050.1 | XP_011522352.1 | 10 | 2790 | |||
XM_011524051.2 | XP_011522353.1 | 12 | 1885 | |||
XM_011524052.2 | XP_011522354.1 | 11 | 1999 | |||
XM_011524053.2 | XP_011522355.1 | 12 | 1912 | |||
XM_011524054.2 | XP_011522356.1 | 12 | 1879 | |||
XM_011524055.2 | XP_011522357.1 | 10 | 1806 | |||
XM_011524056.2 | XP_011522358.1 | 7 | 1759 | |||
XM_011524057.2 | XP_011522359.1 | 11 | 1988 | |||
XM_011524058.2 | XP_011522360.1 | 11 | 1988 | |||
XM_011524059.2 | XP_011522361.1 | 11 | 1988 | |||
XM_017025290.1 | XP_016880779.1 | 9 | 2628 | |||
GenBank mRNA | AB449915.1 | - | 5 | 342 | ||
AY143550.1 | - | 6 | 2041 | |||
BX647719.1 | - | 14 | 2573 | |||
X63546.1 | - | 6 | 2038 | |||
X63547.1 | - | 6 | 2038 |
Gene Symbol: TBC1D3J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025494.1 | XP_016880983.1 | 4 | 261 |
Gene Symbol: TBC1D3P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033892.1 | 6 | 1133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
