siRNA Details
Gene Symbol: USP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304284.1 | NP_001291213.1 | 18 | 2976 | ||
NM_004505.3 | NP_004496.2 | 10 | 2445 | |||
XM_011524050.1 | XP_011522352.1 | 14 | 3194 | |||
XM_011524051.2 | XP_011522353.1 | 16 | 2289 | |||
XM_011524052.2 | XP_011522354.1 | 15 | 2403 | |||
XM_011524053.2 | XP_011522355.1 | 16 | 2316 | |||
XM_011524054.2 | XP_011522356.1 | 16 | 2283 | |||
XM_011524055.2 | XP_011522357.1 | 14 | 2210 | |||
XM_011524056.2 | XP_011522358.1 | 11 | 2163 | |||
XM_011524057.2 | XP_011522359.1 | 15 | 2392 | |||
XM_011524058.2 | XP_011522360.1 | 15 | 2392 | |||
XM_011524059.2 | XP_011522361.1 | 15 | 2392 | |||
XM_017025290.1 | XP_016880779.1 | 13 | 3032 | |||
GenBank mRNA | AB449915.1 | - | 9 | 746 | ||
AY143550.1 | - | 10 | 2445 | |||
BX647719.1 | - | 18 | 2977 | |||
X63546.1 | - | 10 | 2442 | |||
X63547.1 | - | 10 | 2442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
