siRNA Details
Gene Symbol: TBC1D3P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033892.1 | 8, 9 | 1345 |
Gene Symbol: TBC1D29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015594.2 | NP_056409.1 | 1 | 77 | ||
GenBank mRNA | AL117394.1 | - | 2 | 2426 | ||
BC096718.1 | - | 1 | 77 | 40003088 | ||
BC098126.1 | - | 1 | 77 | 40003085 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
