siRNA Details
Gene Symbol: CCL3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001437.3 | NP_001001437.2 | 3 | 539 | ||
GenBank mRNA | BC007783.2 | - | 3 | 534 | 4308472 | |
BC146919.1 | - | 3 | 500 | 9056847 | ||
BC147008.1 | - | 3 | 500 | 9056936 | ||
BC147013.1 | - | 3 | 500 | 9056941 | ||
CF128666.1 | - | 3 | 518 |
Gene Symbol: CCL3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021006.5 | NP_066286.1 | 3 | 549 | ||
NR_111964.1 | 3 | 741 | ||||
GenBank mRNA | BC027888.1 | - | 3 | 537 | 5222247 | |
BC107710.1 | - | 3 | 553 | 6699133 | ||
BC146914.1 | - | 3 | 500 | 9056842 | ||
X52149.1 | - | 3 | 727 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
