siRNA Details
Gene Symbol: CCL3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021006.5 | NP_066286.1 | 1 | 12 | ||
NR_111964.1 | 1 | 12 | ||||
GenBank mRNA | AV739733.1 | - | 56 | |||
BC107710.1 | - | 1 | 16 | 6699133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
