siRNA Details
Gene Symbol: CCL3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001437.3 | NP_001001437.2 | 1 | 49 | ||
GenBank mRNA | AA380745.1 | - | 1 | 49 | ||
BC007783.2 | - | 1 | 44 | 4308472 | ||
CF128666.1 | - | 1 | 28 |
Gene Symbol: CCL3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021006.5 | NP_066286.1 | 1 | 59 | ||
NR_111964.1 | 1 | 59 | ||||
GenBank mRNA | AV739733.1 | - | 1 | 103 | ||
BC027888.1 | - | 1 | 47 | 5222247 | ||
BC107710.1 | - | 1 | 63 | 6699133 | ||
X52149.1 | - | 1 | 45 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
