siRNA Details
Gene Symbol: CCL3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001437.3 | NP_001001437.2 | 3 | 291 | ||
GenBank mRNA | AI302038.1 | - | 3 | 192 | ||
BC007783.2 | - | 3 | 286 | 4308472 | ||
BC146919.1 | - | 3 | 252 | 9056847 | ||
BC147008.1 | - | 3 | 252 | 9056936 | ||
BC147013.1 | - | 3 | 252 | 9056941 | ||
BT006783.1 | - | 3 | 216 | |||
CF128666.1 | - | 3 | 270 |
Gene Symbol: CCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | D63785.1 | - | 3 | 196 |
Gene Symbol: CCL3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021006.5 | NP_066286.1 | 3 | 301 | ||
NR_111964.1 | 3 | 493 | ||||
GenBank mRNA | BC027888.1 | - | 3 | 289 | 5222247 | |
BC107710.1 | - | 3 | 305 | 6699133 | ||
BC146914.1 | - | 3 | 252 | 9056842 | ||
X52149.1 | - | 3 | 479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
