siRNA Details
Gene Symbol: DEFB107B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040705.1 | NP_001035795.1 | 2 | 270 |
Gene Symbol: DEFB107A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037668.1 | NP_001032757.2 | 2 | 270 | ||
GenBank mRNA | AF540979.1 | - | 2 | 170 | ||
AY122467.1 | - | 2 | 170 | |||
BC137510.1 | - | 2 | 196 | 9021510 | ||
BC137513.1 | - | 2 | 196 | 9021513 | ||
DQ119826.1 | - | 2 | 270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
