siRNA Details
Gene Symbol: BCAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170714.1 | NP_001164185.1 | 3 | 532 | ||
NM_001170715.1 | NP_001164186.1 | 2 | 237 | |||
NM_001170716.1 | NP_001164187.1 | 2 | 194 | |||
NM_001170717.1 | NP_001164188.1 | 2 | 256 | |||
NM_001170718.1 | NP_001164189.1 | 2 | 281 | |||
NM_001170719.1 | NP_001164190.1 | 2 | 371 | |||
NM_001170720.1 | NP_001164191.1 | 3 | 386 | |||
NM_014567.3 | NP_055382.2 | 2 | 256 | |||
XM_005256258.4 | XP_005256315.1 | 2 | 456 | |||
XM_005256259.4 | XP_005256316.1 | 2 | 3368 | |||
XM_005256260.4 | XP_005256317.1 | 2 | 377 | |||
XM_011523475.2 | XP_011521777.1 | 2 | 405 | |||
XM_017023897.1 | XP_016879386.1 | 2 | 963 | |||
XM_017023898.1 | XP_016879387.1 | 2 | 963 | |||
XM_017023899.1 | XP_016879388.1 | 2 | 963 | |||
GenBank mRNA | AB040024.1 | - | 2 | 251 | ||
AF218451.1 | - | 2 | 234 | |||
AJ242987.2 | - | 2 | 231 | |||
AK027608.1 | - | 2 | 281 | |||
AK124526.1 | - | 2 | 1676 | |||
AK124815.1 | - | 2 | 240 | |||
AK293808.1 | - | 3 | 532 | |||
AK294513.1 | - | 2 | 287 | |||
AK295809.1 | - | 2 | 166 | |||
AK302617.1 | - | 3 | 386 | |||
AK314722.1 | - | 2 | 256 | |||
AY545071.1 | - | 1 | 110 | |||
BC062556.1 | - | 1 | 111 | 6427808 | ||
DA418642.1 | - | 2 | 256 | |||
DC310094.1 | - | 2 | 371 | |||
DC317947.1 | - | 2 | 197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
