siRNA Details
Gene Symbol: DEFB106A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152251.3 | NP_689464.1 | 11 | |||
GenBank mRNA | AY122466.1 | - | 38 | |||
BC100844.2 | - | 11 | 40002124 | |||
BC100845.2 | - | 11 | 40002125 | |||
BC100846.2 | - | 11 | 40002126 | |||
BC100847.2 | - | 11 | 40002135 | |||
BC110062.1 | - | 11 | 40002127 | |||
BC111688.1 | - | 11 | 40002128 |
Gene Symbol: DEFB106B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040704.1 | NP_001035794.1 | 11 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
