siRNA Details
Gene Symbol: SPATA31A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145196.1 | NP_001138668.1 | 3 | 316 | ||
XM_011517871.2 | XP_011516173.1 | 3 | 323 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
