siRNA Details
Gene Symbol: SSX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278697.1 | NP_001265626.1 | 5, 6 | 473 | ||
NM_003147.5 | NP_003138.3 | 5, 6 | 473 | |||
NM_175698.2 | NP_783629.1 | 5, 6 | 473 | |||
GenBank mRNA | BC002818.2 | - | 5, 6 | 407 | 3636268 | |
BC007343.2 | - | 5, 6 | 407 | 3615133 | ||
BC016957.1 | - | 5, 6 | 407 | 3916128 | ||
BC069313.1 | - | 4, 5 | 337 | 7262129 | ||
BC071827.1 | - | 5, 6 | 407 | 4641362 | ||
BC103863.2 | - | 5, 6 | 475 | 40003337 | ||
BG104299.1 | - | 5, 6 | 444 | |||
BI560469.1 | - | 5, 6 | 479 | |||
JX014236.1 | - | 4, 5 | 337 | |||
X86175.1 | - | 5, 6 | 428 | |||
Z49105.1 | - | 5, 6 | 435 |
Gene Symbol: SSX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278691.1 | NP_001265620.1 | 6, 7 | 559 | ||
NM_005635.3 | NP_005626.1 | 5, 6 | 473 | |||
GenBank mRNA | BC001003.2 | - | 5, 6 | 418 | 3445470 | |
BC125151.1 | - | 5, 6 | 416 | 40120554 | ||
BC128611.1 | - | 5, 6 | 415 | 6159943 | ||
BC133693.1 | - | 5, 6 | 416 | 40120550 | ||
BC150487.2 | - | 5, 6 | 420 | 40120555 | ||
BE777476.1 | - | 6, 7 | 494 | |||
BF211314.1 | - | 5, 6 | 437 | |||
BI831262.1 | - | 5, 6 | 471 | |||
X86174.1 | - | 5, 6 | 428 |
Gene Symbol: SSX2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164417.2 | NP_001157889.1 | 5, 6 | 472 | ||
NM_001278701.1 | NP_001265630.1 | 5, 6 | 472 | |||
NM_001278702.1 | NP_001265631.1 | 5, 6 | 472 | |||
GenBank mRNA | BE391023.1 | - | 5, 6 | 426 | ||
CB158955.1 | - | 5, 6 | 389 |
Gene Symbol: SSX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021014.3 | NP_066294.1 | 5, 6 | 477 | ||
XM_011543885.2 | XP_011542187.1 | 5, 6 | 430 | |||
GenBank mRNA | AK310513.1 | - | 5, 6 | 430 | ||
BC005904.1 | - | 5, 6 | 431 | 4064072 | ||
BC103862.1 | - | 5, 6 | 476 | 40003336 | ||
S82471.1 | - | 4, 5 | 337 | |||
U90840.1 | - | 5, 6 | 390 |
Gene Symbol: SSX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005636.3 | NP_005627.1 | 5, 6 | 395 | ||
XM_017029759.1 | XP_016885248.1 | 5, 6 | 453 | |||
GenBank mRNA | AX740218.1 | - | 4, 5 | 337 | ||
BC005325.1 | - | 5, 6 | 395 | 3961688 | ||
BC137394.1 | - | 4, 5 | 362 | 9021392 | ||
BC137395.1 | - | 4, 5 | 362 | 9021393 | ||
U90841.1 | - | 4, 5 | 337 |
Gene Symbol: SSX4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034832.3 | NP_001030004.1 | 5, 6 | 406 | ||
XM_017029613.1 | XP_016885102.1 | 5, 6 | 453 |
Gene Symbol: SSX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173358.2 | NP_775494.1 | 5, 6 | 496 | ||
XM_017029430.1 | XP_016884919.1 | 4, 5 | 337 |
Gene Symbol: SSX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027250.2 | 5, 6 | 621 | |||
GenBank mRNA | AK302291.1 | - | 6, 7 | 825 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
