siRNA Details
Gene Symbol: SSX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173358.2 | NP_775494.1 | 8 | 771 | ||
XM_017029430.1 | XP_016884919.1 | 8 | 717 | |||
XM_017029431.1 | XP_016884920.1 | 6 | 476 |
Gene Symbol: SSX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278691.1 | NP_001265620.1 | 9 | 834 | ||
NM_005635.3 | NP_005626.1 | 8 | 748 | |||
GenBank mRNA | BC001003.2 | - | 8 | 693 | 3445470 | |
BC125151.1 | - | 8 | 691 | 40120554 | ||
BC128611.1 | - | 8 | 690 | 6159943 | ||
BC133693.1 | - | 8 | 691 | 40120550 | ||
BC150487.2 | - | 8 | 695 | 40120555 | ||
X86174.1 | - | 8 | 703 |
Gene Symbol: SSX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278697.1 | NP_001265626.1 | 9 | 826 | ||
NM_003147.5 | NP_003138.3 | 9 | 894 | |||
NM_175698.2 | NP_783629.1 | 8 | 748 | |||
GenBank mRNA | BC002818.2 | - | 9 | 828 | 3636268 | |
BC007343.2 | - | 8 | 682 | 3615133 | ||
BC016957.1 | - | 8 | 682 | 3916128 | ||
BC071827.1 | - | 9 | 828 | 4641362 | ||
BC103863.2 | - | 8 | 750 | 40003337 | ||
BI560469.1 | - | 8 | 756 | |||
X86175.1 | - | 8 | 703 | |||
Z49105.1 | - | 8 | 710 |
Gene Symbol: SSX2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164417.2 | NP_001157889.1 | 8 | 747 | ||
NM_001278701.1 | NP_001265630.1 | 9 | 893 | |||
NM_001278702.1 | NP_001265631.1 | 9 | 825 |
Gene Symbol: SSX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021014.3 | NP_066294.1 | 8 | 752 | ||
GenBank mRNA | BC103862.1 | - | 8 | 751 | 40003336 | |
S82471.1 | - | 7 | 612 | |||
U90840.1 | - | 8 | 665 |
Gene Symbol: SSX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005636.3 | NP_005627.1 | 8 | 670 | ||
NM_175729.1 | NP_783856.1 | 7 | 534 | |||
XM_017029759.1 | XP_016885248.1 | 8 | 728 | |||
GenBank mRNA | BC005325.1 | - | 8 | 670 | 3961688 |
Gene Symbol: SSX4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034832.3 | NP_001030004.1 | 8 | 681 | ||
NM_001040612.2 | NP_001035702.1 | 7 | 545 | |||
XM_017029613.1 | XP_016885102.1 | 8 | 728 |
Gene Symbol: SSX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021015.3 | NP_066295.3 | 9 | 788 | ||
NM_175723.1 | NP_783729.1 | 8 | 665 | |||
GenBank mRNA | BC016640.2 | - | 8 | 754 | 3923927 |
Gene Symbol: SSX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028366.1 | 8 | 740 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
