siRNA Details
Gene Symbol: XAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017523.3 | NP_059993.2 | 2, 3 | 418 | ||
NR_046396.1 | 2, 3 | 418 | ||||
XM_005256706.4 | XP_005256763.1 | 2, 3 | 990 | |||
XM_005256708.2 | XP_005256765.1 | 2, 3 | 843 | |||
XM_011523945.2 | XP_011522247.1 | 3, 4 | 805 | |||
XM_011523946.2 | XP_011522248.1 | 3, 4 | 805 | |||
XM_011523947.2 | XP_011522249.1 | 2, 3 | 326 | |||
XM_011523948.1 | XP_011522250.1 | 3, 4 | 805 | |||
GenBank mRNA | AK290276.1 | - | 3, 4 | 351 | ||
AK292710.1 | - | 2, 3 | 198 | |||
BC073156.1 | - | 2, 3 | 217 | 6068243 | ||
BM475564.1 | - | 2, 3 | 189 | |||
EF028165.1 | - | 2, 3 | 176 | |||
X99699.1 | - | 2, 3 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
