siRNA Details
Gene Symbol: RBMY1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320944.1 | NP_001307873.1 | 11 | 1616 | ||
NM_001320945.1 | NP_001307874.1 | 11 | 1549 | |||
NM_005058.3 | NP_005049.1 | 12 | 1727 | |||
XM_011531438.1 | XP_011529740.1 | 9 | 1333 | |||
XM_017030060.1 | XP_016885549.1 | 10 | 1392 | |||
GenBank mRNA | BC047768.1 | - | 11 | 1524 | 5744153 | |
BC070298.1 | - | 11 | 1581 | 6615329 | ||
BX119894.1 | - | 1 | 211 | |||
X76059.1 | - | 12 | 1692 |
Gene Symbol: RBMY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011531433.1 | XP_011529735.1 | 9 | 1354 | ||
XM_011531434.1 | XP_011529736.1 | 11 | 1526 | |||
XM_017030057.1 | XP_016885546.1 | 10 | 1392 |
Gene Symbol: RBMY1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006120.3 | NP_001006120.2 | 12 | 1727 | ||
NM_001320949.1 | NP_001307878.1 | 11 | 1616 | |||
NM_001320950.1 | NP_001307879.1 | 12 | 1667 | |||
XM_011531486.2 | XP_011529788.1 | 12 | 1600 | |||
XM_011531489.1 | XP_011529791.1 | 9 | 1354 | |||
XM_011531490.1 | XP_011529792.1 | 11 | 1526 |
Gene Symbol: RBMY1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006118.2 | NP_001006118.2 | 12 | 1687 | ||
XM_011531493.1 | XP_011529795.1 | 11 | 1576 | |||
XM_011531495.1 | XP_011529797.1 | 9 | 1354 | |||
XM_011531496.1 | XP_011529798.1 | 11 | 1526 | |||
XM_017030058.1 | XP_016885547.1 | 10 | 1392 |
Gene Symbol: RBMY1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303410.1 | NP_001290339.1 | 12 | 1682 | ||
NM_152585.2 | NP_689798.1 | 12 | 1687 | |||
XM_017030033.1 | XP_016885522.1 | 10 | 1392 | |||
GenBank mRNA | BC030018.2 | - | 12 | 1687 | 5269012 | |
X76060.1 | - | 12 | 1664 |
Gene Symbol: RBMY1J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011531500.1 | XP_011529802.1 | 11 | 1512 | ||
XM_011531502.1 | XP_011529804.1 | 9 | 1290 | |||
XM_011531503.1 | XP_011529805.1 | 9 | 1276 | |||
XM_017030059.1 | XP_016885548.1 | 10 | 1392 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
