siRNA Details
Gene Symbol: LOC101929950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006722258.3 | XP_006722321.1 | 4 | 478 | ||
XM_017025500.1 | XP_016880989.1 | 4 | 483 |
Gene Symbol: LOC440434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036750.2 | 5 | 676 |
Gene Symbol: NPEPPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006310.3 | NP_006301.3 | 5 | 858 | ||
XM_011525496.1 | XP_011523798.1 | 6 | 660 | |||
XM_017025372.1 | XP_016880861.1 | 5 | 820 | |||
XM_017025373.1 | XP_016880862.1 | 6 | 660 | |||
GenBank mRNA | AJ132583.1 | - | 5 | 588 | ||
AK095877.1 | - | 6 | 624 | |||
AK096709.1 | - | 7 | 645 | |||
AK297107.1 | - | 4 | 362 | |||
AK303037.1 | - | 6 | 653 | |||
AK311414.1 | - | 449 | ||||
BC065294.1 | - | 5 | 547 | 6059589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
