siRNA Details
Gene Symbol: NCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000265.5 | NP_000256.4 | 2 | 170 | ||
GenBank mRNA | AK127905.1 | - | 2 | 121 | ||
AK223217.1 | - | 2 | 121 | |||
AK292094.1 | - | 2 | 121 | |||
AK316341.1 | - | 2 | 121 | |||
AK316552.1 | - | 2 | 121 | |||
BC002816.2 | - | 2 | 170 | 3633829 | ||
BC065731.1 | - | 2 | 126 | 6735930 |
Gene Symbol: NCF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003186.1 | 2 | 105 | |||
GenBank mRNA | AK298655.1 | - | 2 | 121 | ||
BC080194.1 | - | 2 | 105 |
Gene Symbol: NCF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003187.3 | 2 | 153 | |||
GenBank mRNA | AF330627.1 | - | 2 | 110 | ||
AK298739.1 | - | 2 | 119 | |||
AK301397.1 | - | 2 | 120 | |||
AK316329.1 | - | 2 | 153 | |||
DC389713.1 | - | 2 | 153 | |||
M25665.1 | - | 2 | 112 | |||
M55067.1 | - | 2 | 122 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
