siRNA Details
Gene Symbol: NCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000265.5 | NP_000256.4 | 3 | 272 | ||
GenBank mRNA | AK127905.1 | - | 3 | 296 | ||
AK223217.1 | - | 3 | 223 | |||
AK292094.1 | - | 3 | 223 | |||
AK316552.1 | - | 3 | 223 | |||
BC002816.2 | - | 3 | 272 | 3633829 | ||
BC065731.1 | - | 3 | 228 | 6735930 |
Gene Symbol: NCF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK298655.1 | - | 3 | 223 |
Gene Symbol: NCF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003187.3 | 3 | 255 | |||
GenBank mRNA | AF330627.1 | - | 3 | 212 | ||
AK316329.1 | - | 3 | 255 | |||
DC389713.1 | - | 3 | 255 | |||
M25665.1 | - | 3 | 214 | |||
M55067.1 | - | 3 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
