siRNA Details
Gene Symbol: SRSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191005.2 | NP_001177934.1 | 4 | 624 | ||
NM_001191006.2 | NP_001177935.1 | 4 | 624 | |||
NM_001191007.2 | NP_001177936.1 | 4 | 624 | |||
NM_001191009.2 | NP_001177938.1 | 4 | 624 | |||
NM_001300936.1 | NP_001287865.1 | 4 | 624 | |||
NM_001300937.1 | NP_001287866.1 | 4 | 624 | |||
NM_006625.5 | NP_006616.1 | 4 | 624 | |||
NM_054016.3 | NP_473357.1 | 4 | 624 | |||
NR_034035.2 | 5 | 1021 | ||||
XM_017000100.1 | XP_016855589.1 | 4 | 4344 | |||
XM_017000101.1 | XP_016855590.1 | 5 | 3887 | |||
XM_017000102.1 | XP_016855591.1 | 5 | 1514 | |||
GenBank mRNA | AF047448.1 | - | 4 | 445 | ||
AF067730.1 | - | 4 | 445 | |||
AF419331.1 | - | 4 | 492 | |||
AF449427.1 | - | 4 | 416 | |||
AK001286.1 | - | 4 | 488 | |||
AK001656.1 | - | 4 | 488 | |||
AK055311.1 | - | 2 | 1426 | |||
AK093226.1 | - | 5 | 1485 | |||
AK125834.1 | - | 2 | 2821 | |||
AK222994.1 | - | 4 | 515 | |||
AK296175.1 | - | 4 | 517 | |||
AY007101.1 | - | 4 | 457 | |||
AY150180.1 | - | 4 | 430 | |||
AY150181.1 | - | 4 | 430 | |||
BC001107.1 | - | 4 | 517 | 3504780 | ||
BC005039.1 | - | 4 | 480 | 3531441 | ||
BC010074.2 | - | 4 | 469 | 4302658 | ||
BC071575.1 | - | 4 | 3745 | |||
BX648939.1 | - | 1 | 5641 | |||
KU178536.1 | - | 4 | 416 | |||
KU178537.1 | - | 2 | 207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
