siRNA Details
Gene Symbol: ANKRD20A11P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027270.1 | 1 | 633 | |||
GenBank mRNA | AF426257.1 | - | 1 | 633 | ||
BC043199.1 | - | 1 | 633 |
Gene Symbol: ANKRD20A5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040113.1 | 1 | 391 | |||
GenBank mRNA | BC022023.1 | - | 1 | 391 | 4824658 | |
BC127726.1 | - | 1 | 170 | 40127384 | ||
BP201899.1 | - | 1 | 119 | |||
DB446908.1 | - | 1 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
