siRNA Details
Gene Symbol: ANKRD20A5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040113.1 | 1 | 473 | |||
GenBank mRNA | BC022023.1 | - | 1 | 472 | 4824658 | |
BC127726.1 | - | 1 | 252 | 40127384 | ||
BP201899.1 | - | 1 | 201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
