siRNA Details
Gene Symbol: HSFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033108.2 | NP_149099.2 | 1 | 117 | ||
NM_152584.1 | NP_689797.1 | 1 | 114 | |||
NR_003510.1 | 1 | 114 | ||||
XM_005262565.3 | XP_005262622.1 | 1 | 86 | |||
GenBank mRNA | AF332226.1 | - | 1 | 114 | ||
AF332227.1 | - | 1 | 114 | |||
AJ566404.1 | - | 1 | 114 | |||
BC036567.2 | - | 1 | 117 | 5297256 | ||
BC055414.1 | - | 1 | 89 | 6616624 | ||
BC117380.1 | - | 1 | 58 | 40125931 | ||
BC117382.1 | - | 1 | 58 | 40125933 |
Gene Symbol: HSFY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001877.1 | NP_001001877.1 | 1 | 114 | ||
NM_153716.1 | NP_714927.1 | 1 | 134 | |||
NR_003509.1 | 1 | 134 | ||||
XM_005262508.3 | XP_005262565.1 | 1 | 134 | |||
XM_017030031.1 | XP_016885520.1 | 1 | 113 | |||
GenBank mRNA | AK058182.1 | - | 1 | 134 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
