siRNA Details
Gene Symbol: KPNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320611.1 | NP_001307540.1 | 7 | 1323 | ||
NM_002266.3 | NP_002257.1 | 7 | 965 | |||
GenBank mRNA | AK024833.1 | - | 7 | 833 | ||
AK058111.1 | - | 7 | 834 | |||
AK291954.1 | - | 7 | 832 | |||
BC005978.1 | - | 7 | 832 | 4105058 | ||
BC013257.1 | - | 7 | 828 | |||
BC034443.1 | - | 7 | 791 | |||
BC053343.1 | - | 7 | 821 | 4450470 | ||
BC067848.1 | - | 7 | 1064 | 4825235 | ||
BC146905.1 | - | 7 | 819 | 9056833 | ||
BT006665.1 | - | 6 | 703 | |||
U09559.1 | - | 7 | 835 | |||
U28386.1 | - | 7 | 835 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
