siRNA Details
Gene Symbol: TAS2R20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176889.2 | NP_795370.2 | 1 | 586 | ||
GenBank mRNA | BC100915.2 | - | 1 | 586 | 40003843 | |
BC100916.2 | - | 1 | 586 | 40003844 | ||
BC100917.2 | - | 1 | 586 | 40003847 | ||
BC100918.2 | - | 1 | 586 | 40003851 | ||
BC113843.1 | - | 1 | 586 | 40003846 | ||
BC114451.1 | - | 1 | 586 | 40003845 |
Gene Symbol: TAS2R30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097643.1 | NP_001091112.1 | 1 | 586 |
Gene Symbol: TAS2R46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176887.2 | NP_795368.2 | 1 | 586 | ||
GenBank mRNA | BC115401.2 | - | 1 | 586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
