siRNA Details
Gene Symbol: RGPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078170.2 | NP_001071638.2 | 5 | 641 | ||
XM_017004845.1 | XP_016860334.1 | 9 | 1092 | |||
XM_017004846.1 | XP_016860335.1 | 5 | 763 | |||
XM_017004847.1 | XP_016860336.1 | 5 | 760 | |||
XM_017004848.1 | XP_016860337.1 | 9 | 1092 | |||
XM_017004849.1 | XP_016860338.1 | 9 | 1092 | |||
XM_017004850.1 | XP_016860339.1 | 9 | 1092 | |||
GenBank mRNA | AK302429.1 | - | 5 | 641 |
Gene Symbol: RGPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024457.3 | NP_001019628.3 | 5 | 595 | ||
XM_011532845.2 | XP_011531147.1 | 6 | 794 | |||
XM_011532849.2 | XP_011531151.1 | 6 | 794 | |||
XM_011532850.1 | XP_011531152.1 | 6 | 794 | |||
XM_011532851.2 | XP_011531153.1 | 6 | 794 | |||
XM_011532852.2 | XP_011531154.1 | 6 | 794 | |||
XM_017004103.1 | XP_016859592.1 | 6 | 794 | |||
XM_017004104.1 | XP_016859593.1 | 5 | 911 | |||
XM_017004105.1 | XP_016859594.1 | 5 | 759 | |||
XM_017004106.1 | XP_016859595.1 | 5 | 878 | |||
XM_017004107.1 | XP_016859596.1 | 6 | 794 | |||
GenBank mRNA | CR749330.1 | - | 5 | 595 |
Gene Symbol: RGPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144013.1 | NP_001137485.1 | 5 | 663 | ||
XM_017004738.1 | XP_016860227.1 | 6 | 712 | |||
XM_017004739.1 | XP_016860228.1 | 6 | 712 | |||
XM_017004740.1 | XP_016860229.1 | 5 | 637 |
Gene Symbol: RGPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182588.2 | NP_872394.2 | 5 | 688 | ||
XM_011511017.2 | XP_011509319.1 | 5 | 635 | |||
XM_011511018.2 | XP_011509320.1 | 5 | 638 | |||
XM_011511019.2 | XP_011509321.1 | 5 | 634 | |||
XM_011511020.2 | XP_011509322.1 | 5 | 634 | |||
XM_017003898.1 | XP_016859387.1 | 5 | 699 | |||
GenBank mRNA | BX537861.1 | - | 4 | 559 |
Gene Symbol: RGPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005054.2 | NP_005045.2 | 5 | 683 | ||
NM_032260.2 | NP_115636.1 | 6 | 1139 | |||
XM_006712794.3 | XP_006712857.2 | 5 | 714 | |||
XM_017005080.1 | XP_016860569.1 | 5 | 703 | |||
XM_017005081.1 | XP_016860570.1 | 5 | 714 | |||
XM_017005082.1 | XP_016860571.1 | 4 | 531 | |||
GenBank mRNA | AL136868.1 | - | 6 | 1140 | ||
EU794682.1 | - | 5 | 683 |
Gene Symbol: RGPD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037866.1 | NP_001032955.1 | 6 | 1139 | ||
NM_001123363.3 | NP_001116835.1 | 6 | 1139 | |||
XM_011511766.2 | XP_011510068.1 | 5 | 723 | |||
XM_011511767.2 | XP_011510069.1 | 5 | 723 | |||
XM_011511769.2 | XP_011510071.1 | 5 | 723 | |||
XM_011511770.2 | XP_011510072.1 | 5 | 723 | |||
XM_017004839.1 | XP_016860328.1 | 5 | 723 | |||
XM_017004840.1 | XP_016860329.1 | 5 | 796 | |||
XM_017004841.1 | XP_016860330.1 | 5 | 926 | |||
XM_017004842.1 | XP_016860331.1 | 5 | 703 | |||
XM_017004843.1 | XP_016860332.1 | 5 | 723 | |||
XM_017004844.1 | XP_016860333.1 | 4 | 531 | |||
GenBank mRNA | AK056675.1 | - | 2 | 268 |
Gene Symbol: RGPD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164463.1 | NP_001157935.1 | 5 | 798 | ||
XM_005263747.4 | XP_005263804.1 | 5 | 796 | |||
XM_011511733.2 | XP_011510035.1 | 5 | 796 | |||
XM_017004825.1 | XP_016860314.1 | 5 | 796 | |||
XM_017004826.1 | XP_016860315.1 | 5 | 796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
