siRNA Details
Gene Symbol: RGPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078170.2 | NP_001071638.2 | 1, 2 | 116 | ||
XM_017004846.1 | XP_016860335.1 | 1, 2 | 238 | |||
GenBank mRNA | AK302429.1 | - | 1, 2 | 116 |
Gene Symbol: RGPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017004105.1 | XP_016859594.1 | 1, 2 | 234 | ||
XM_017004106.1 | XP_016859595.1 | 1, 2 | 353 | |||
GenBank mRNA | DC343949.1 | - | 1, 2 | 145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
