siRNA Details
Gene Symbol: NPIPB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310148.1 | NP_001297077.1 | 6 | 816 | ||
XM_017023244.1 | XP_016878733.1 | 5 | 2827 | |||
XM_017023245.1 | XP_016878734.1 | 6 | 2664 | |||
XM_017023246.1 | XP_016878735.1 | 6 | 2585 | |||
XM_017023247.1 | XP_016878736.1 | 5 | 818 |
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130464.2 | NP_569731.2 | 6 | 667 | ||
GenBank mRNA | AB209632.1 | - | 5 | 3234 | ||
AK294452.1 | - | 2 | 230 | |||
AK295170.1 | - | 6 | 666 | |||
AK303500.1 | - | 2 | 161 | |||
AK304162.1 | - | 4 | 449 | |||
AK308629.1 | - | 1 | 210 | |||
AK316459.1 | - | 2 | 96 | |||
BC036263.1 | - | 6 | 667 | 3903371 | ||
BC094882.1 | - | 5 | 594 | 3093162 | ||
BM557638.1 | - | 2 | 241 | |||
KF175175.1 | - | 5 | 534 | |||
KF175176.1 | - | 5 | 591 | |||
KF175177.1 | - | 5 | 591 | |||
KF175178.1 | - | 5 | 492 | |||
KF175179.1 | - | 5 | 534 | |||
KF175180.1 | - | 5 | 534 | |||
KF175181.1 | - | 5 | 678 | |||
KF175182.1 | - | 5 | 534 | |||
KF175183.1 | - | 4 | 426 | |||
KF175184.1 | - | 4 | 417 | |||
KF175185.1 | - | 5 | 534 | |||
KF175186.1 | - | 4 | 408 | |||
KF175187.1 | - | 5 | 534 | |||
KF175188.1 | - | 5 | 495 | |||
KF175189.1 | - | 5 | 495 | |||
KF175190.1 | - | 5 | 534 | |||
KF175191.1 | - | 5 | 495 | |||
KF175192.1 | - | 5 | 495 | |||
KF175193.1 | - | 5 | 498 | |||
KF175194.1 | - | 5 | 498 | |||
KF175197.1 | - | 5 | 495 | |||
KF175198.1 | - | 5 | 591 | |||
KF175199.1 | - | 5 | 591 | |||
KF175200.1 | - | 5 | 591 | |||
KF175201.1 | - | 5 | 534 | |||
KF175202.1 | - | 5 | 495 | |||
KF175203.1 | - | 5 | 591 | |||
KF175215.1 | - | 7 | 853 | |||
KF175216.1 | - | 6 | 868 | |||
KF175223.1 | - | 5 | 591 | |||
KF175224.1 | - | 5 | 534 |
Gene Symbol: NPIPB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135865.1 | NP_001129337.1 | 5 | 870 | ||
GenBank mRNA | AK296338.1 | - | 5 | 711 | ||
AK303264.1 | - | 2 | 183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
