siRNA Details
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098845.2 | NP_001092315.2 | 6 | 738 | ||
GenBank mRNA | AK223149.1 | - | 6 | 552 | ||
AK313626.1 | - | 6 | 601 | |||
X16662.1 | - | 6 | 582 |
Gene Symbol: ANXA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040084.2 | NP_001035173.1 | 6 | 701 | ||
NM_001271702.1 | NP_001258631.1 | 6 | 815 | |||
NM_001271703.1 | NP_001258632.1 | 4 | 515 | |||
XM_006717951.3 | XP_006718014.1 | 6 | 655 | |||
XM_011540096.1 | XP_011538398.1 | 7 | 854 | |||
XM_011540098.1 | XP_011538400.1 | 7 | 854 | |||
XM_011540099.1 | XP_011538401.1 | 7 | 854 | |||
GenBank mRNA | AK296573.1 | - | 4 | 366 | ||
AK298761.1 | - | 6 | 681 | |||
AK300139.1 | - | 6 | 666 | |||
BC004376.1 | - | 6 | 598 | 3639064 | ||
BC073755.1 | - | 6 | 632 | 6538697 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
