siRNA Details
Gene Symbol: ANXA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040084.2 | NP_001035173.1 | 5 | 580 | ||
NM_001271702.1 | NP_001258631.1 | 5 | 694 | |||
NM_001271703.1 | NP_001258632.1 | 3 | 394 | |||
XM_006717951.3 | XP_006718014.1 | 5 | 534 | |||
XM_011540096.1 | XP_011538398.1 | 6 | 733 | |||
XM_011540098.1 | XP_011538400.1 | 6 | 733 | |||
XM_011540099.1 | XP_011538401.1 | 6 | 733 | |||
XM_011540101.2 | XP_011538403.1 | 6 | 733 | |||
GenBank mRNA | AK296573.1 | - | 3 | 245 | ||
AK298761.1 | - | 5 | 560 | |||
AK300139.1 | - | 5 | 545 | |||
AK300158.1 | - | 5 | 438 | |||
BC004376.1 | - | 5 | 477 | 3639064 | ||
BC073755.1 | - | 5 | 511 | 6538697 | ||
M81844.1 | - | 5 | 382 |
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098845.2 | NP_001092315.2 | 5 | 617 | ||
GenBank mRNA | AK223149.1 | - | 5 | 431 | ||
AK313626.1 | - | 5 | 480 | |||
X16662.1 | - | 5 | 461 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
