siRNA Details
Gene Symbol: ZNF761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008401.3 | NP_001008401.3 | 5 | 558 | ||
NM_001289951.1 | NP_001276880.1 | 4 | 339 | |||
NM_001289952.1 | NP_001276881.1 | 4 | 407 | |||
NM_001289953.1 | NP_001276882.1 | 3 | 284 | |||
NR_110551.1 | 5 | 462 | ||||
GenBank mRNA | AB107355.2 | - | 4 | 408 | ||
BC071639.1 | - | 2 | 132 | |||
BC103708.1 | - | 5 | 443 | |||
DA237674.1 | - | 5 | 558 | |||
DB232487.1 | - | 4 | 331 |
Gene Symbol: ZNF525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003699.1 | 3 | 185 | |||
GenBank mRNA | AK097037.1 | - | 3 | 205 | ||
DB221179.1 | - | 3 | 185 |
Gene Symbol: ZNF765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040185.1 | NP_001035275.1 | 3 | 223 | ||
GenBank mRNA | AK293890.1 | - | 3 | 222 | ||
AK316300.1 | - | 3 | 217 | |||
BC001610.1 | - | 2 | 185 | |||
BC017357.1 | - | 2 | 186 | |||
BC140724.1 | - | 3 | 197 | 9021592 | ||
BX647981.1 | - | 3 | 215 | |||
CD656338.1 | - | 3 | 244 |
Gene Symbol: ZNF813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004301.3 | NP_001004301.2 | 3 | 228 | ||
GenBank mRNA | DB235327.1 | - | 3 | 219 |
Gene Symbol: ZNF83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DB254966.1 | - | 5 | 429 |
Gene Symbol: ZNF845 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321522.1 | NP_001308451.1 | 4 | 402 | ||
NM_001321523.1 | NP_001308452.1 | 4 | 341 | |||
NM_001321524.1 | NP_001308453.1 | 4 | 349 | |||
NM_138374.2 | NP_612383.1 | 3 | 247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
