siRNA Details
Gene Symbol: ZNF665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024733.3 | NP_079009.3 | 3 | 225 | ||
XM_005259266.4 | XP_005259323.1 | 3 | 259 | |||
XM_011527325.2 | XP_011525627.1 | 3 | 330 | |||
XM_011527327.2 | XP_011525629.1 | 4 | 634 | |||
GenBank mRNA | AK291403.1 | - | 3 | 215 | ||
AL832507.1 | - | 3 | 210 |
Gene Symbol: ZNF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006969.3 | NP_008900.3 | 3 | 282 | ||
NR_036599.1 | 3 | 282 | ||||
XM_006723354.3 | XP_006723417.1 | 3 | 299 | |||
XM_011527262.2 | XP_011525564.1 | 3 | 312 | |||
GenBank mRNA | AK304704.1 | - | 3 | 246 | ||
BC070146.1 | - | 3 | 246 | |||
BC140720.1 | - | 1 | 65 | 9021588 | ||
BX485233.1 | - | 3 | 294 | |||
BX489697.1 | - | 3 | 245 | |||
CN263784.1 | - | 3 | 229 | |||
CR933598.1 | - | 3 | 245 |
Gene Symbol: ZNF320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207333.2 | NP_997216.2 | 3 | 316 | ||
XM_006723059.3 | XP_006723122.1 | 4 | 362 | |||
XM_017026393.1 | XP_016881882.1 | 5 | 607 | |||
XM_017026394.1 | XP_016881883.1 | 5 | 532 | |||
XM_017026395.1 | XP_016881884.1 | 5 | 680 | |||
XM_017026396.1 | XP_016881885.1 | 4 | 449 | |||
XM_017026397.1 | XP_016881886.1 | 4 | 454 | |||
XM_017026398.1 | XP_016881887.1 | 4 | 333 | |||
XM_017026399.1 | XP_016881888.1 | 3 | 326 | |||
XM_017026400.1 | XP_016881889.1 | 3 | 246 | |||
GenBank mRNA | AK307375.1 | - | 4 | 363 | ||
BC064408.1 | - | 4 | 543 | |||
BC105738.1 | - | 7 | 628 | |||
BC131555.1 | - | 2 | 171 | 40108031 | ||
CR933622.1 | - | 1 | 103 |
Gene Symbol: ZNF525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003699.1 | 3 | 210 | |||
GenBank mRNA | AK097037.1 | - | 3 | 230 | ||
DB221179.1 | - | 3 | 210 |
Gene Symbol: ZNF528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032423.2 | NP_115799.2 | 5 | 551 | ||
XM_005259323.1 | XP_005259380.1 | 1 | 33 | |||
XM_006723418.2 | XP_006723481.1 | 5 | 547 | |||
XM_011527392.1 | XP_011525694.1 | 5 | 543 | |||
XM_017027360.1 | XP_016882849.1 | 6 | 664 | |||
XM_017027361.1 | XP_016882850.1 | 6 | 660 | |||
XM_017027362.1 | XP_016882851.1 | 6 | 711 | |||
XM_017027363.1 | XP_016882852.1 | 7 | 705 | |||
XM_017027364.1 | XP_016882853.1 | 6 | 660 | |||
GenBank mRNA | AK056552.1 | - | 5 | 618 | ||
AL833325.1 | - | 5 | 551 | |||
BC101712.1 | - | 2 | 167 | 8069218 | ||
BC111950.1 | - | 2 | 179 | 8327418 |
Gene Symbol: ZNF701 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172655.1 | NP_001166126.1 | 4 | 548 | ||
NM_018260.2 | NP_060730.2 | 3 | 254 | |||
XM_011527092.1 | XP_011525394.1 | 3 | 328 | |||
GenBank mRNA | AK001753.1 | - | 3 | 252 | ||
BC054884.1 | - | 3 | 222 | |||
BC067346.1 | - | 4 | 548 | |||
BC080612.1 | - | 2 | 210 | 5816672 | ||
BC131719.1 | - | 3 | 228 | 40123413 | ||
BC136425.1 | - | 3 | 358 | 9020414 | ||
BC136426.1 | - | 3 | 226 | 9020415 |
Gene Symbol: ZNF761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008401.3 | NP_001008401.3 | 5 | 583 | ||
NM_001289951.1 | NP_001276880.1 | 4 | 364 | |||
NM_001289952.1 | NP_001276881.1 | 4 | 432 | |||
NM_001289953.1 | NP_001276882.1 | 3 | 309 | |||
NR_110551.1 | 5 | 487 | ||||
GenBank mRNA | AB107355.2 | - | 4 | 433 | ||
BC071639.1 | - | 2 | 157 | |||
BC103708.1 | - | 5 | 468 | |||
DB232487.1 | - | 4 | 356 |
Gene Symbol: ZNF765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040185.1 | NP_001035275.1 | 3 | 248 | ||
GenBank mRNA | AK293890.1 | - | 3 | 247 | ||
AK316300.1 | - | 3 | 242 | |||
BC001610.1 | - | 2 | 210 | |||
BC017357.1 | - | 2 | 211 | |||
BC140724.1 | - | 3 | 222 | 9021592 | ||
BX647981.1 | - | 3 | 240 | |||
CD656338.1 | - | 3 | 269 |
Gene Symbol: ZNF813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004301.3 | NP_001004301.2 | 3 | 253 | ||
GenBank mRNA | DB235327.1 | - | 3 | 244 | ||
DR006226.1 | - | 1 | 73 |
Gene Symbol: ZNF83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105549.1 | NP_001099019.1 | 3 | 278 | ||
NM_001105550.1 | NP_001099020.1 | 3 | 278 | |||
NM_001105551.1 | NP_001099021.1 | 3 | 278 | |||
NM_001105552.1 | NP_001099022.1 | 3 | 278 | |||
NM_001277945.1 | NP_001264874.1 | 5 | 546 | |||
NM_001277948.1 | NP_001264877.1 | 4 | 419 | |||
NM_001277949.1 | NP_001264878.1 | 4 | 458 | |||
NM_001277952.1 | NP_001264881.1 | 3 | 278 | |||
XM_017026951.1 | XP_016882440.1 | 5 | 524 | |||
GenBank mRNA | AK075066.1 | - | 5 | 453 | ||
BC032867.1 | - | 3 | 239 | 5264072 | ||
BX641726.1 | - | 3 | 238 | |||
DA151844.1 | - | 3 | 235 | |||
DA183557.1 | - | 3 | 278 | |||
DA258790.1 | - | 3 | 233 | |||
DA341245.1 | - | 3 | 250 | |||
DA434207.1 | - | 4 | 363 | |||
DA449506.1 | - | 3 | 243 | |||
DA497861.1 | - | 3 | 302 | |||
DB134270.1 | - | 4 | 329 | |||
DB252431.1 | - | 3 | 252 | |||
DB254966.1 | - | 5 | 454 | |||
DB272526.1 | - | 4 | 329 | |||
DT216882.1 | - | 6 | 532 |
Gene Symbol: ZNF845 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321522.1 | NP_001308451.1 | 4 | 427 | ||
NM_001321523.1 | NP_001308452.1 | 4 | 366 | |||
NM_001321524.1 | NP_001308453.1 | 4 | 374 | |||
NM_138374.2 | NP_612383.1 | 3 | 272 | |||
GenBank mRNA | AK295079.1 | - | 4 | 346 | ||
DC400704.1 | - | 3 | 244 |
Gene Symbol: ZNF888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310127.1 | NP_001297056.1 | 2 | 183 | ||
XM_005259451.4 | XP_005259508.1 | 4 | 415 | |||
XM_017026798.1 | XP_016882287.1 | 4 | 576 | |||
XM_017026799.1 | XP_016882288.1 | 4 | 461 | |||
XM_017026800.1 | XP_016882289.1 | 2 | 192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
