siRNA Details
Gene Symbol: AMZ2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026903.1 | 5 | 1747 | |||
GenBank mRNA | AK056627.1 | - | 5 | 1730 | ||
AK311317.1 | - | 4 | 466 | |||
BC073870.1 | - | 5 | 816 | |||
BX537538.1 | - | 4 | 1935 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
