siRNA Details
Gene Symbol: AMZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033569.1 | NP_001028741.1 | 5 | 675 | ||
NM_001033570.1 | NP_001028742.1 | 5 | 671 | |||
NM_001033571.1 | NP_001028743.1 | 5 | 647 | |||
NM_001033572.1 | NP_001028744.1 | 5 | 643 | |||
NM_001033574.1 | NP_001028746.1 | 3 | 1054 | |||
NM_001289054.1 | NP_001275983.1 | 5 | 655 | |||
NM_001289056.1 | NP_001275985.1 | 5 | 746 | |||
NM_016627.4 | NP_057711.3 | 4 | 1228 | |||
XM_005257432.4 | XP_005257489.1 | 4 | 1279 | |||
XM_005257433.3 | XP_005257490.1 | 5 | 677 | |||
XM_005257435.4 | XP_005257492.1 | 5 | 619 | |||
XM_005257436.3 | XP_005257493.1 | 5 | 1006 | |||
XM_006721940.2 | XP_006722003.1 | 5 | 617 | |||
XM_017024723.1 | XP_016880212.1 | 5 | 866 | |||
XM_017024724.1 | XP_016880213.1 | 5 | 615 | |||
XM_017024725.1 | XP_016880214.1 | 4 | 1191 | |||
XM_017024726.1 | XP_016880215.1 | 5 | 652 | |||
XM_017024727.1 | XP_016880216.1 | 5 | 653 | |||
XM_017024728.1 | XP_016880217.1 | 4 | 720 | |||
GenBank mRNA | AJ635358.1 | - | 3 | 526 | ||
AK090981.1 | - | 3 | 1514 | |||
AK126146.1 | - | 4 | 1658 | |||
BC050709.1 | - | 5 | 607 | 6060235 | ||
BC056271.1 | - | 5 | 640 | 4102397 | ||
BM450564.1 | - | 5 | 616 | |||
BQ432895.1 | - | 3 | 515 | |||
CN278874.1 | - | 5 | 644 |
Gene Symbol: AMZ2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026903.1 | 4 | 1550 | |||
GenBank mRNA | AK056627.1 | - | 4 | 1533 | ||
AK311317.1 | - | 3 | 269 | |||
BC073870.1 | - | 4 | 619 | |||
BX537538.1 | - | 3 | 1738 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
