siRNA Details
Gene Symbol: AMZ2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026903.1 | 4 | 1573 | |||
GenBank mRNA | AK056627.1 | - | 4 | 1556 | ||
AK311317.1 | - | 3 | 292 | |||
BC073870.1 | - | 4 | 642 | |||
BX537538.1 | - | 3 | 1761 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
