siRNA Details
Gene Symbol: FAM25A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146157.2 | NP_001139629.1 | 3 | 191 | ||
GenBank mRNA | BC171878.1 | - | 3 | 181 | 9054532 | |
BC171915.1 | - | 3 | 181 | 9054569 |
Gene Symbol: FAM25E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017042.1 | XP_016872531.1 | 2 | 1953 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
