siRNA Details
Gene Symbol: FAM25C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137548.2 | NP_001131020.1 | 3 | 255 | ||
GenBank mRNA | BC146955.1 | - | 3 | 252 | 9056883 | |
BC146972.1 | - | 3 | 255 | 9056900 | ||
BC146989.1 | - | 3 | 255 | 9056917 | ||
BC146990.1 | - | 3 | 255 | 9056918 | ||
BC146991.1 | - | 3 | 255 | 9056919 | ||
BC147022.1 | - | 3 | 255 | 9056950 | ||
BC147026.1 | - | 3 | 255 | 9056954 | ||
BC171871.1 | - | 3 | 255 | 9054525 | ||
BX093303.1 | - | 3 | 310 |
Gene Symbol: FAM25A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146157.2 | NP_001139629.1 | 3 | 265 | ||
GenBank mRNA | BC171878.1 | - | 3 | 255 | 9054532 | |
BC171915.1 | - | 3 | 255 | 9054569 |
Gene Symbol: FAM25BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104039.1 | 3 | 255 | |||
GenBank mRNA | BC146968.1 | - | 3 | 252 | 9056896 |
Gene Symbol: FAM25E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017042.1 | XP_016872531.1 | 2 | 2027 |
Gene Symbol: FAM25G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137549.1 | NP_001131021.1 | 3 | 255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
