siRNA Details
Gene Symbol: BROX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288579.1 | NP_001275508.1 | 1 | 104 | ||
NM_144695.3 | NP_653296.2 | 1 | 104 | |||
NR_109927.1 | 1 | 104 | ||||
XM_005273069.4 | XP_005273126.1 | 1 | 93 | |||
XM_006711173.3 | XP_006711236.1 | 1 | 93 | |||
XM_017000374.1 | XP_016855863.1 | 1 | 93 | |||
XM_017000376.1 | XP_016855865.1 | 1 | 93 | |||
GenBank mRNA | AK056983.1 | - | 1 | 93 | ||
AL701416.1 | - | 1 | 112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
