siRNA Details
Gene Symbol: BROX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288579.1 | NP_001275508.1 | 1, 2 | 391 | ||
NM_144695.3 | NP_653296.2 | 1, 2 | 391 | |||
GenBank mRNA | AK056983.1 | - | 1, 2 | 380 | ||
BC113635.1 | - | 1, 2 | 128 | 8322687 | ||
BC113637.1 | - | 1, 2 | 128 | 8322689 | ||
BC143811.1 | - | 1, 2 | 128 | 9052326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
