siRNA Details
Gene Symbol: BROX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288579.1 | NP_001275508.1 | 7 | 928 | ||
NM_001288580.1 | NP_001275509.1 | 7 | 634 | |||
NM_001288581.1 | NP_001275510.1 | 8 | 730 | |||
NM_144695.3 | NP_653296.2 | 8 | 1024 | |||
XM_005273065.2 | XP_005273122.2 | 8 | 660 | |||
XM_005273069.4 | XP_005273126.1 | 7 | 896 | |||
XM_006711173.3 | XP_006711236.1 | 9 | 1093 | |||
XM_011509212.2 | XP_011507514.1 | 7 | 987 | |||
XM_011509213.2 | XP_011507515.1 | 7 | 968 | |||
XM_011509214.2 | XP_011507516.1 | 9 | 893 | |||
XM_017000374.1 | XP_016855863.1 | 10 | 1218 | |||
XM_017000375.1 | XP_016855864.1 | 6 | 873 | |||
XM_017000376.1 | XP_016855865.1 | 8 | 1021 | |||
GenBank mRNA | AB276123.1 | - | 7 | 618 | ||
AK056983.1 | - | 8 | 1013 | |||
AK293974.1 | - | 7 | 560 | |||
AK304445.1 | - | 8 | 675 | |||
AK315930.1 | - | 7 | 564 | |||
BC113635.1 | - | 8 | 761 | 8322687 | ||
BC113637.1 | - | 8 | 761 | 8322689 | ||
BC143811.1 | - | 7 | 665 | 9052326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
