siRNA Details
Gene Symbol: LOC101059949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030105.1 | XP_016885594.1 | 1 | 756 |
Gene Symbol: ANKRD20A12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046228.1 | 3 | 1056 | |||
GenBank mRNA | AK090412.1 | - | 3 | 2057 |
Gene Symbol: LOC101927345 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544009.2 | XP_011542311.1 | 1 | 279 |
Gene Symbol: LOC105379272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544006.2 | XP_011542308.1 | 2 | 400 |
Gene Symbol: LOC644249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015390.1 | XP_016870879.1 | 3 | 467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
