siRNA Details
Gene Symbol: ANKRD18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147195.2 | NP_671728.2 | 14 | 2713 | ||
XM_006716749.3 | XP_006716812.1 | 15 | 2990 | |||
XM_011517836.2 | XP_011516138.1 | 15 | 2991 | |||
XM_011517837.2 | XP_011516139.1 | 15 | 2991 | |||
XM_011517839.2 | XP_011516141.1 | 14 | 2935 | |||
XM_017014576.1 | XP_016870065.1 | 16 | 3122 | |||
GenBank mRNA | AB095935.1 | - | 14 | 2658 | ||
AK093059.1 | - | 1 | 49 | |||
BC131497.1 | - | 1 | 45 | |||
BC152434.1 | - | 14 | 2681 | 8862553 | ||
DB066861.1 | - | 1 | 102 |
Gene Symbol: ANKRD18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244752.1 | NP_001231681.1 | 14 | 2736 | ||
XM_011517881.2 | XP_011516183.1 | 15 | 2803 | |||
XM_011517883.2 | XP_011516185.1 | 14 | 2746 | |||
XM_011517885.2 | XP_011516187.1 | 9 | 1891 | |||
XM_017014728.1 | XP_016870217.1 | 9 | 1888 |
Gene Symbol: ANKRD18CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136286.1 | 16 | 2900 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
