siRNA Details
Gene Symbol: LOC100133331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028327.1 | 3 | 3664 | |||
GenBank mRNA | AK097080.1 | - | 4 | 1812 |
Gene Symbol: LINC00265 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026999.1 | 12 | 4165 | |||
GenBank mRNA | AK097404.1 | - | 2 | 1709 |
Gene Symbol: LINC00999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024497.2 | 8 | 4672 | |||
GenBank mRNA | AK128746.1 | - | 2 | 3131 |
Gene Symbol: LINC01000 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024368.1 | 5 | 3774 | |||
GenBank mRNA | AK294679.1 | - | 1 | 1502 | ||
BC047304.1 | - | 1 | 744 | |||
BC062355.1 | - | 6 | 2220 | |||
BC131589.1 | - | 6 | 2212 |
Gene Symbol: LINC01001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028326.1 | 3 | 4005 |
Gene Symbol: LINC01002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028324.1 | 3 | 4376 | |||
GenBank mRNA | AK128780.1 | - | 2 | 1681 | ||
BC117566.1 | - | 1 | 427 |
Gene Symbol: LINC01061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037596.1 | 3 | 4213 |
Gene Symbol: LOC100132062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028325.1 | 3 | 3761 | |||
GenBank mRNA | BC112924.1 | - | 2 | 2112 |
Gene Symbol: LOC100132217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128836.1 | - | 3 | 3417 |
Gene Symbol: LOC100132287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028322.1 | 3 | 3761 |
Gene Symbol: LOC107986805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012932.1 | XP_016868421.1 | 1 | 1633 |
Gene Symbol: LOC401357 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130727.1 | 1 | 3137 | |||
GenBank mRNA | AK125790.1 | - | 1 | 3136 | ||
AK128114.1 | - | 1 | 2878 |
Gene Symbol: LOC729737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039983.2 | 3 | 4865 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
