siRNA Details
Gene Symbol: LOC100133331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028327.1 | 3 | 3746 | |||
GenBank mRNA | AK097080.1 | - | 4 | 1894 |
Gene Symbol: LINC00999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128746.1 | - | 2 | 3218 |
Gene Symbol: LINC01000 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024368.1 | 5 | 3856 | |||
GenBank mRNA | AK093729.1 | - | 1 | 736 | ||
AK294679.1 | - | 1 | 1584 | |||
BC047304.1 | - | 1 | 826 | |||
BC062355.1 | - | 6 | 2302 |
Gene Symbol: LINC01001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028326.1 | 3 | 4087 |
Gene Symbol: LINC01002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC117566.1 | - | 1 | 509 |
Gene Symbol: LOC100132062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028325.1 | 3 | 3843 | |||
GenBank mRNA | BC112924.1 | - | 2 | 2194 |
Gene Symbol: LOC100132287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028322.1 | 3 | 3843 |
Gene Symbol: LOC729737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039983.2 | 3 | 4947 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
