siRNA Details
Gene Symbol: SPRR2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006945.4 | NP_008876.3 | 2 | 306 | ||
XM_017002175.1 | XP_016857664.1 | 2 | 338 | |||
GenBank mRNA | AK291317.1 | - | 2 | 308 | ||
BC093801.1 | - | 2 | 290 | |||
BC093803.1 | - | 2 | 291 | |||
BC120938.1 | - | 2 | 290 | 40115065 | ||
BG183546.1 | - | 3 | 535 |
Gene Symbol: SPRR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017418.2 | NP_001017418.1 | 2 | 309 | ||
XM_017002174.1 | XP_016857663.1 | 2 | 305 | |||
GenBank mRNA | BF090370.1 | - | 1 | 181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
