siRNA Details
Gene Symbol: SPRR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005988.2 | NP_005979.1 | 1, 2 | 44 | ||
GenBank mRNA | AK311939.1 | - | 1, 2 | 45 | ||
BC096108.1 | - | 17 | 40002232 | |||
BC096109.1 | - | 17 | 40002234 | |||
BC096110.1 | - | 17 | 40002235 | |||
BC096111.1 | - | 17 | 40002236 | |||
BC128049.1 | - | 1, 2 | 27 | 40115066 | ||
BE710686.1 | - | 263 |
Gene Symbol: SPRR2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006945.4 | NP_008876.3 | 1, 2 | 43 | ||
GenBank mRNA | AK291317.1 | - | 1, 2 | 45 | ||
BC093801.1 | - | 1, 2 | 27 | |||
BC093803.1 | - | 1, 2 | 28 | |||
BC120938.1 | - | 1, 2 | 27 | 40115065 | ||
M21302.1 | - | 1, 2 | 43 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
